4-Amino-2,3,5-trichlorpyridin
Lieferant: CHEMPUR
Synonyme:
2,3,5-trichloropyridin-4-amine
RJC01063.250
RJC01063.1
CHMPRJC01063.250EA
0
EUR
CHMPRJC01063.250
MAYBRJC01063.1
4-Amino-2,3,5-trichlorpyridin
4-Amino-2,3,5-trichloropyridine
Formel:
C₅H₃Cl₃N₂ Molecular Weight: 197.45 g/mol |
MDL:
MFCD00153908 CAS-Nummer: 28443-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...