3-Amino-5-chlor-2-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-5-chloropicoline, 5-Chloro-2-methylpyridin-3-amine

PY-1687-1G PY-1687-5G
COBBPY-1687-1GEA 274 EUR
COBBPY-1687-1G COBBPY-1687-5G
3-Amino-5-chlor-2-methylpyridin
3-Amino-5-chloro-2-methylpyridine
Formel: C₆H₇ClN₂
Molecular Weight: 142,5862 g/mol
MDL: MFCD12196974
CAS-Nummer: 89639-36-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR