4,4'-Diacetylbiphenyl ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15911.06
A15911.14
A15911.06EA
81.5
EUR
A15911.06
A15911.14
4,4'-Diacetylbiphenyl ≥98%
4,4'-Diacetylbiphenyl
Purity: 98%
CAS No.: 787-69-9
Molecular Formula: C16H14O2
Formula Weight: 238.29
Melting Point: 190-193°
MDL No.: MFCD00017248
Formel:
C₁₆H₁₄O₂ Molecular Weight: 238.29 g/mol Schmelzpunkt: 190…193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017248 CAS-Nummer: 787-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...