2-(2,2,2-Trifluorethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(2,2,2-Trifluoroethoxy)phenyl)boronic acid, 2-(2,2,2-Trifluoroethoxy)benzeneboronic acid

BB-3851-5G BB-3851-1G
COBBBB-3851-5GEA 884 EUR
COBBBB-3851-5G COBBBB-3851-1G
2-(2,2,2-Trifluorethoxy)phenylboronsäure
2-(2,2,2-Trifluorethoxy)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219,95 g/mol
MDL: MFCD09027258
CAS-Nummer: 957060-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR