2-Fluor-3-(trifluormethoxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5449-5G
BB-5449-1G
COBBBB-5449-5GEA
0
EUR
COBBBB-5449-5G
COBBBB-5449-1G
2-Fluor-3-(trifluormethoxy)phenylboronsäure
2-Fluor-3-(trifluormethoxy)phenylboronsäure
Formel:
C₇H₅BF₄O₃ |
MDL:
MFCD11045053 CAS-Nummer: 881402-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...