2-Fluor-5-(trifluormethoxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Fluoro-5-(trifluoromethoxy)phenyl)boronic acid
BB-3580-5G
BB-3580-1G
COBBBB-3580-5GEA
312
EUR
COBBBB-3580-5G
COBBBB-3580-1G
2-Fluor-5-(trifluormethoxy)phenylboronsäure
2-Fluor-5-(trifluormethoxy)phenylboronsäure
Formel:
C₇H₅BF₄O₃ Molecular Weight: 223.9200 g/mol |
MDL:
MFCD09027234 CAS-Nummer: 881402-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...