3-Fluor-4-(trifluormethoxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Fluoro-4-(trifluoromethoxy)phenyl)boronic acid
BB-9047-5G
BB-9047-1G
COBBBB-9047-5GEA
720
EUR
COBBBB-9047-5G
COBBBB-9047-1G
3-Fluor-4-(trifluormethoxy)phenylboronsäure
3-Fluor-4-(trifluormethoxy)phenylboronsäure
Formel:
C₇H₅BF₄O₃ Molecular Weight: 223.920 g/mol |
MDL:
MFCD07368680 CAS-Nummer: 187804-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...