4-Fluor-3-(2,2,2-trifluorethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Fluoro-3-(2,2,2-trifluoroethoxy)phenyl)boronic acid

BB-3941-5G BB-3941-25G BB-3941-1G
COBBBB-3941-5GEA 0 EUR
COBBBB-3941-5G COBBBB-3941-25G COBBBB-3941-1G
4-Fluor-3-(2,2,2-trifluorethoxy)phenylboronsäure
4-Fluor-3-(2,2,2-trifluorethoxy)phenylboronsäure
Formel: C₈H₇BF₄O₃
MDL: MFCD09475813
CAS-Nummer: 957034-62-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR