Tris(2,4,6-trimethylphenyl)phosphin ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme: Nalpha-Acetyl-D-arginine, Trimesitylphosphine
B21116.03 B21116.06
B21116.03EA 75.2 EUR
B21116.03 B21116.06
Tris(2,4,6-trimethylphenyl)phosphin ≥98%
Tris(2,4,6-trimethylphenyl)phosphin
Purity: 98% CAS No.: 23897-15-6 Molecular Formula: C27H33P Formula Weight: 388.54 Melting Point: 185-188° MDL No.: MFCD00014912
Formel: C₂₇H₃₃P
Molecular Weight: 388.54 g/mol
Schmelzpunkt: 185…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014912
CAS-Nummer: 23897-15-6