2,4-Dichlor-6-methylbenzonitril
Lieferant: Thermo Scientific
TL00275.1
TL00275.10
TL00275.50
MAYBTL00275.1EA
0
EUR
MAYBTL00275.1
MAYBTL00275.10
MAYBTL00275.50
2,4-Dichlor-6-methylbenzonitril
2,4-Dichlor-6-methylbenzonitril
Formel:
C₈H₅Cl₂N Molecular Weight: 186,04 g/mol |
MDL:
MFCD00052865 CAS-Nummer: 175277-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...