3,6-Dibrompicolinsäure

Lieferant: COMBI-BLOCKS

PY-7889-1G PY-7889-5G
COBBPY-7889-1GEA 170 EUR
COBBPY-7889-1G COBBPY-7889-5G
3,6-Dibrompicolinsäure
3,6-Dibrompicolinsäure
Formel: C₆H₃Br₂NO₂
Molecular Weight: 280,9015 g/mol
MDL: MFCD11855997
CAS-Nummer: 1133116-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR