4-Brom-3-picolin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-3-methylpyridine hydrochloride

OS-7578-1G OS-7578-25G OS-7578-5G
COBBOS-7578-1GEA 149 EUR
COBBOS-7578-1G COBBOS-7578-25G COBBOS-7578-5G
4-Brom-3-picolin Hydrochlorid
4-Brom-3-picolin Hydrochlorid
Formel: C₆H₇BrClN
Molecular Weight: 208.483 g/mol
MDL: MFCD04966778
CAS-Nummer: 40899-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR