trans-3,4,4-Trimethyl-2-pentensäure
Lieferant: COMBI-BLOCKS
HC-2929-1G
HC-2929-5G
COBBHC-2929-1GEA
204
EUR
COBBHC-2929-1G
COBBHC-2929-5G
trans-3,4,4-Trimethyl-2-pentensäure
trans-3,4,4-Trimethyl-2-pentenoic acid
Formel:
C₈H₁₄O₂ |
MDL:
MFCD11172805 CAS-Nummer: 99799-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...