1H,1H,7H-Dodecafluorheptylacrylat

Lieferant: FLUOROCHEM

Synonyme: DFHA, Acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl ester, 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Acrylate

006058-5G 006058-25G
FLUO006058-5GEA 110 EUR
FLUO006058-5G FLUO006058-25G
1H,1H,7H-Dodecafluorheptylacrylat
1H,1H,7H-Dodecafluorheptylacrylat
Formel: C₁₀H₆F₁₂O₂
Molecular Weight: 386.14 g/mol
MDL: MFCD00080746
CAS-Nummer: 2993-85-3
EINECS: 221-064-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR