Tetraethylammoniumtrifluormethansulfonat

Lieferant: CHEMPUR

Synonyme: Tetraethylammoniumtrifluoromethanesulphonate

FL9431.POR 009431-1G 009431-5G
CHMPFL9431.POREA 0 EUR
CHMPFL9431.POR FLUO009431-1G FLUO009431-5G
Tetraethylammoniumtrifluormethansulfonat
Tetraethylammoniumtrifluormethansulfonat
Formel: C₉H₂₀F₃NO₃S
MDL: MFCD00042586
CAS-Nummer: 35895-69-3
EINECS: 252-781-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR