3,4-Dinitrofluorbenzol

Lieferant: CHEMPUR

Synonyme: 4-Fluoro-1,2-dinitrobenzene

FL4965.POR 004965-25G 004965-5G
CHMPFL4965.POREA 0 EUR
CHMPFL4965.POR FLUO004965-25G FLUO004965-5G
3,4-Dinitrofluorbenzol
3,4-Dinitrofluorbenzol
Formel: C₆H₃FN₂O₄
Molecular Weight: 186,1 g/mol
MDL: MFCD00236671
CAS-Nummer: 364-53-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR