2-(Trifluormethoxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4,4,5,5-Tetramethyl-2-(2-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane

PN-2650-1G PN-2650-5G
COBBPN-2650-1GEA 0 EUR
COBBPN-2650-1G COBBPN-2650-5G
2-(Trifluormethoxy)phenylboronsäurepinakolester
2-(Trifluormethoxy)phenylboronsäurepinakolester
Formel: C₁₃H₁₆BF₃O₃
MDL: MFCD05863918
CAS-Nummer: 832114-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR