4-(Trifluormethoxy)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4,4,5,5-Tetramethyl-2-(4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane
PN-2709-25G
PN-2709-5G
PN-2709-1G
COBBPN-2709-25GEA
884
EUR
COBBPN-2709-25G
COBBPN-2709-5G
COBBPN-2709-1G
4-(Trifluormethoxy)phenylboronsäurepinakolester
4-(Trifluormethoxy)phenylboronsäurepinakolester
Formel:
C₁₃H₁₆BF₃O₃ Molecular Weight: 288,0706 g/mol |
MDL:
MFCD05863920 CAS-Nummer: 474709-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...