2-Brom-5-(trifluormethoxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-3944-5G
BB-3944-1G
COBBBB-3944-5GEA
0
EUR
COBBBB-3944-5G
COBBBB-3944-1G
2-Brom-5-(trifluormethoxy)phenylboronsäure
2-Brom-5-(trifluormethoxy)phenylboronsäure
Formel:
C₇H₅BBrF₃O₃ |
MDL:
MFCD09475807 CAS-Nummer: 957034-55-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...