4-(Propylthio)phenylboronsäure
Lieferant: COMBI-BLOCKS
FA-1340-1G
FA-1340-25G
FA-1340-5G
COBBFA-1340-1GEA
208
EUR
COBBFA-1340-1G
COBBFA-1340-25G
COBBFA-1340-5G
4-(Propylthio)phenylboronsäure
4-(Propylthio)phenylboronsäure
Formel:
C₉H₁₃BO₂S Molecular Weight: 196,074 g/mol |
MDL:
MFCD06801730 CAS-Nummer: 279261-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...