2-(Trifluormethyl)-4-chinolinol

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluoromethyl)quinolin-4-ol, 4-Hydroxy-2-(trifluoromethyl)quinoline

CFF290.10 CFF290.50 007672-1G 007672-5G
CHMPCFF290.10EA 532 EUR
CHMPCFF290.10 CHMPCFF290.50 FLUO007672-1G FLUO007672-5G
2-(Trifluormethyl)-4-chinolinol
2-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,1560 g/mol
MDL: MFCD00153196
CAS-Nummer: 1701-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR