6-(Trifluormethyl)-4-chinolinol

Lieferant: FLUOROCHEM

Synonyme: 4-Hydroxy-6-(trifluoromethyl)quinoline

CFF1182.1 007673-1G 007673-5G
CHMPCFF1182.1EA 522 EUR
CHMPCFF1182.1 FLUO007673-1G FLUO007673-5G
6-(Trifluormethyl)-4-chinolinol
6-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,16 g/mol
MDL: MFCD00209673
CAS-Nummer: 49713-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR