5-Isopropyl-1,3-cyclohexandion
Lieferant: FLUOROCHEM
Synonyme:
5-Isopropylcyclohexane-1,3-dione
011178-5G
011178-1G
FLUO011178-5GEA
806
EUR
FLUO011178-5G
FLUO011178-1G
5-Isopropyl-1,3-cyclohexandion
5-Isopropyl-1,3-cyclohexandion
Formel:
C₉H₁₄O₂·H₂O Molecular Weight: 154,21 g/mol (anhy) |
MDL:
MFCD00075263 CAS-Nummer: 18456-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...