Ethyl-3-brom-6-chlorpicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
Ethyl 3-bromo-6-chloropicolinate
PY-7780-1G
PY-7780-5G
COBBPY-7780-1GEA
154
EUR
COBBPY-7780-1G
COBBPY-7780-5G
Ethyl-3-brom-6-chlorpicolinat
Ethyl-3-brom-6-chlorpicolinat
Formel:
C₈H₇BrClNO₂ Molecular Weight: 264,5037 g/mol |
MDL:
MFCD11053875 CAS-Nummer: 1065074-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...