Ethyl-3,6-dibrompicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 3,6-dibromopicolinate

PY-1246-5G PY-1246-1G
COBBPY-1246-5GEA 560 EUR
COBBPY-1246-5G COBBPY-1246-1G
Ethyl-3,6-dibrompicolinat
Ethyl-3,6-dibrompicolinat
Formel: C₈H₇Br₂NO₂
Molecular Weight: 308,9600 g/mol
MDL: MFCD13195784
CAS-Nummer: 1214375-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR