Methyl-3,6-dibrompicolinat
Lieferant: COMBI-BLOCKS
PY-7376-1G
PY-7376-5G
COBBPY-7376-1GEA
303
EUR
COBBPY-7376-1G
COBBPY-7376-5G
Methyl-3,6-dibrompicolinat
Methyl-3,6-dibrompicolinat
Formel:
C₇H₅Br₂NO₂ |
MDL:
MFCD10699713 CAS-Nummer: 495416-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...