Methyl-4-brompicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-bromopicolinate, Methyl 4-bromopyridine-2-carboxylate
CA-4118-5G
CA-4118-1G
COBBCA-4118-5GEA
115
EUR
COBBCA-4118-5G
COBBCA-4118-1G
Methyl-4-brompicolinat
Methyl-4-brompicolinat
Formel:
C₇H₆BrNO₂ Molecular Weight: 216,03 g/mol |
MDL:
MFCD07374897 CAS-Nummer: 29681-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...