Ethyl-5-brompicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 5-bromopicolinate, Ethyl 5-bromopyridine-2-carboxylate

PY-7196-1G PY-7196-5G
COBBPY-7196-1GEA 93.7 EUR
COBBPY-7196-1G COBBPY-7196-5G
Ethyl-5-brompicolinat
Ethyl-5-brompicolinat
Formel: C₈H₈BrNO₂
Molecular Weight: 230,0586 g/mol
MDL: MFCD00234415
CAS-Nummer: 77199-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR