Methyl-6-brompicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-bromopicolinate, Methyl 6-bromopyridine-2-carboxylate, 6-Bromopyridine-2-carboxylic acid methyl ester, 6-Bromopicolinic acid methyl ester

CA-4199-1G CA-4199-25G CA-4199-5G
COBBCA-4199-1GEA 26.2 EUR
COBBCA-4199-1G COBBCA-4199-25G COBBCA-4199-5G
Methyl-6-brompicolinat
Methyl-6-brompicolinat
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
MDL: MFCD06203934
CAS-Nummer: 26218-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR