Methyl-6-amino-3-brompicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-amino-3-bromopicolinate, Methyl 6-amino-3-bromopyridine-2-carboxylate

CA-4184-5G CA-4184-1G
COBBCA-4184-5GEA 641 EUR
COBBCA-4184-5G COBBCA-4184-1G
Methyl-6-amino-3-brompicolinat
Methyl-6-amino-3-brompicolinat
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,0467 g/mol
MDL: MFCD09800897
CAS-Nummer: 178876-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR