Methyl-6-aminopicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
6-Aminopicolinic acid methyll ester, Methyl 6-aminopicolinate, Methyl 6-Aminopicolinate, Methyl 6-Aminopyridine-2-carboxylate, 6-Aminopyridine-2-carboxylic acid methyll ester
CA-4144-1G
CA-4144-5G
COBBCA-4144-1GEA
25.9
EUR
COBBCA-4144-1G
COBBCA-4144-5G
Methyl-6-aminopicolinat
Methyl-6-aminopicolinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,1500 g/mol |
MDL:
MFCD00233712 CAS-Nummer: 36052-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...