Methyl-4-methoxypicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-methoxypyridine-2-carboxylate

CA-4110-5G CA-4110-1G
COBBCA-4110-5GEA 114 EUR
COBBCA-4110-5G COBBCA-4110-1G
Methyl-4-methoxypicolinat
Methyl-4-methoxypicolinat
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD00956223
CAS-Nummer: 29681-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR