Methyl-3-fluorpicolinat

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-pyridine-2-carboxylic acid methyl ester, Methyl 3-fluoropyridine-2-carboxylate

PY-1396-1G PY-1396-5G
COBBPY-1396-1GEA 102 EUR
COBBPY-1396-1G COBBPY-1396-5G
Methyl-3-fluorpicolinat
Methyl-3-fluorpicolinat
Formel: C₇H₆FNO₂
Molecular Weight: 155,1300 g/mol
MDL: MFCD06659496
CAS-Nummer: 869108-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR