Methyl-4-iodpicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-iodopyridine-2-carboxylate
CA-4128-5G
CA-4128-1G
COBBCA-4128-5GEA
722
EUR
COBBCA-4128-5G
COBBCA-4128-1G
Methyl-4-iodpicolinat
Methyl-4-iodpicolinat
Formel:
C₇H₆INO₂ |
MDL:
MFCD08235100 CAS-Nummer: 380381-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...