Methyl-3-methylpicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 3-Methylpicolinate, Methyl 3-Methylpyridine-2-carboxylate, Methyl 3-methylpicolinate, 3-Methylpyridine-2-carboxylic acid methyll ester, 3-Methylpicolinic acid methyll ester
PY-7288-1G
PY-7288-25G
PY-7288-5G
COBBPY-7288-1GEA
25.9
EUR
COBBPY-7288-1G
COBBPY-7288-25G
COBBPY-7288-5G
Methyl-3-methylpicolinat
Methyl-3-methylpicolinat
Formel:
C₈H₉NO₂ Molecular Weight: 151.1600 g/mol |
MDL:
MFCD03789583 CAS-Nummer: 59718-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...