1-Methoxy-4-nitronaphthalin

Lieferant: COMBI-BLOCKS

AN-1312-5G AN-1312-1G
COBBAN-1312-5GEA 266 EUR
COBBAN-1312-5G COBBAN-1312-1G
1-Methoxy-4-nitronaphthalin
1-Methoxy-4-nitronaphthalene
Formel: C₁₁H₉NO₃
Molecular Weight: 203,1941 g/mol
MDL: MFCD00003926
CAS-Nummer: 4900-63-4
EINECS: 225-528-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR