2,4-Dichlor-3-methylpyridin

Lieferant: COMBI-BLOCKS

PY-7830-1G PY-7830-250M
COBBPY-7830-1GEA 127 EUR
COBBPY-7830-1G COBBPY-7830-250M
2,4-Dichlor-3-methylpyridin
2,4-Dichloro-3-methylpyridine
Formel: C₆H₅Cl₂N
Molecular Weight: 162,0200 g/mol
MDL: MFCD09263623
CAS-Nummer: 132097-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR