4-Chlor-2-picolin

Lieferant: FLUOROCHEM

Synonyme: 4-Chloro-2-methylpyridine

003362-1G 003362-5G
FLUO003362-1GEA 28.5 EUR
FLUO003362-1G FLUO003362-5G
4-Chlor-2-picolin
4-Chlor-2-picolin
Formel: C₆H₆ClN
Molecular Weight: 127,57 g/mol
MDL: MFCD00038833
CAS-Nummer: 3678-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR