3-Methoxy-2-naphthaldehyd

Lieferant: COMBI-BLOCKS

CA-5006-5G CA-5006-1G
COBBCA-5006-5GEA 571 EUR
COBBCA-5006-5G COBBCA-5006-1G
3-Methoxy-2-naphthaldehyd
3-Methoxy-2-naphthaldehyd
Formel: C₁₂H₁₀O₂
Molecular Weight: 186.2066 g/mol
MDL: MFCD13191799
CAS-Nummer: 56679-88-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR