6-Hydroxy-2-naphthaldehyd

Lieferant: BIONET RESEARCH

5Y-5022.1 5Y-5022.10 5Y-5022.5 5Y-5022.500
BIOT5Y-5022.1EA 0 EUR
BIOT5Y-5022.1 BIOT5Y-5022.10 BIOT5Y-5022.5 BIOT5Y-5022.500
6-Hydroxy-2-naphthaldehyd
6-Hydroxy-2-naphthaldehyd
Formel: C₁₁H₈O₂
Molecular Weight: 172.1800 g/mol
MDL: MFCD03427537
CAS-Nummer: 78119-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR