Methyl-6-(hydroxymethyl)picolinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 6-(hydroxymethyl)picolinate, Methyl 6-(hydroxymethyl)pyridine-2-carboxylate
PY-7740-1G
PY-7740-5G
COBBPY-7740-1GEA
50.8
EUR
COBBPY-7740-1G
COBBPY-7740-5G
Methyl-6-(hydroxymethyl)picolinat
Methyl-6-(hydroxymethyl)picolinat
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol |
MDL:
MFCD03840850 CAS-Nummer: 39977-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...