Tetramethylammoniumacetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
245070-25G
245070-5G
SIAL245070-25GEA
117
EUR
SIAL245070-25G
SIAL245070-5G
Tetramethylammoniumacetat, Sigma-Aldrich®
Tetramethylammoniumacetat
Formel:
C₆H₁₅NO₂ Molecular Weight: 133,20 g/mol |
MDL:
MFCD00011630 CAS-Nummer: 10581-12-1 EINECS: 234-179-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...