(4-Methylphenylthio)aceton

Lieferant: FLUOROCHEM

003767-1G 003767-5G
FLUO003767-1GEA 42.9 EUR
FLUO003767-1G FLUO003767-5G
(4-Methylphenylthio)aceton
(4-Methylphenylthio)aceton
Formel: C₁₀H₁₂OS
Molecular Weight: 180,27 g/mol
MDL: MFCD00052713
CAS-Nummer: 1200-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR