3-(Trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2652-100G BB-2652-5G BB-2652-25G
COBBBB-2652-100GEA 0 EUR
COBBBB-2652-100G COBBBB-2652-5G COBBBB-2652-25G
3-(Trifluormethoxy)phenylboronsäure
3-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
MDL: MFCD01320697
CAS-Nummer: 179113-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR