4-(Trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(4-(trifluormethoxy)phenyl)boran, 4-(Trifluormethoxy)benzolboronsäure, (4-(Trifluormethoxy)phenyl)metaborsäure

BB-2709-5G BB-2709-25G BB-2709-100G
COBBBB-2709-5GEA 37.5 EUR
COBBBB-2709-5G COBBBB-2709-25G COBBBB-2709-100G
4-(Trifluormethoxy)phenylboronsäure
4-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
Molecular Weight: 205.93 g/mol
MDL: MFCD01074648
CAS-Nummer: 139301-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR