3-Brom-2-pyridincarbonitril
Lieferant: COMBI-BLOCKS
OS-7656-1G
OS-7656-25G
OS-7656-5G
COBBOS-7656-1GEA
0
EUR
COBBOS-7656-1G
COBBOS-7656-25G
COBBOS-7656-5G
3-Brom-2-pyridincarbonitril
3-Brom-2-pyridincarbonitril
Formel:
C₆H₃BrN₂ |
MDL:
MFCD02683288 CAS-Nummer: 55758-02-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...