3-Brom-6-fluor-2-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-fluor-6-picolin

OS-7536-5G OS-7536-25G OS-7536-1G
COBBOS-7536-5GEA 38.2 EUR
COBBOS-7536-5G COBBOS-7536-25G COBBOS-7536-1G
3-Brom-6-fluor-2-methylpyridin
3-Brom-6-fluor-2-methylpyridin
Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
MDL: MFCD03095092
CAS-Nummer: 375368-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR