2-Cyan-3-methylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Methyl-2-pyridincarbonitril, 2-Cyan-3-picolin, 2-Cyan-β-picolin
OS-7407-5G
OS-7407-25G
OS-7407-100G
COBBOS-7407-5GEA
25.5
EUR
COBBOS-7407-5G
COBBOS-7407-25G
COBBOS-7407-100G
2-Cyan-3-methylpyridin
2-Cyano-3-methylpyridine
Formel:
C₇H₆N₂ Molecular Weight: 118.14 g/mol Siedepunkt: 139…142 °C (38 mmHg) Schmelzpunkt: 87…90 °C |
MDL:
MFCD00190585 CAS-Nummer: 20970-75-6 EINECS: 244-131-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...