(3-Acetamidophenyl)boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, 3-Acetamidophenylboronic acid Pinacol ester, 3-Acetamidophenylboronic acid, pinacol estet, 2-(3-Acetamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide

PN-2171-5G PN-2171-1G
COBBPN-2171-5GEA 251 EUR
COBBPN-2171-5G COBBPN-2171-1G
(3-Acetamidophenyl)boronsäurepinakolester
(3-Acetamidophenyl)boronsäurepinakolester
Formel: C₁₄H₂₀BNO₃
Molecular Weight: 261.1245 g/mol
MDL: MFCD03789261
CAS-Nummer: 480424-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR