(4-Acetamidophenyl)boronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide, 4-Acetylaminophenylboronic acid pinacol ester, N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide
PN-2124-1G
PN-2124-5G
COBBPN-2124-1GEA
86.2
EUR
COBBPN-2124-1G
COBBPN-2124-5G
(4-Acetamidophenyl)boronsäurepinakolester
(4-Acetamidophenyl)boronsäurepinakolester
Formel:
C₁₄H₂₀BNO₃ Molecular Weight: 261.125 g/mol |
MDL:
MFCD02093722 CAS-Nummer: 214360-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...